PRI424056775 Princeton BioMolecular Research, Inc.

N-{[1-(2,2-difluoroethyl)-5-methyl-1H-pyrazol-4-yl]methyl}cyclopropanamine

  • MDL number MFCD09701767
  • Cas Number : 1170869-32-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart