PRI424056824 Princeton BioMolecular Research, Inc.

1-[4-methyl-6-(trifluoromethyl)pyrimidin-2-yl]piperidin-4-amine

  • MDL number MFCD09701804
  • Cas Number : 1018125-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart