PRI424056835 Princeton BioMolecular Research, Inc.

1-methyl-3-nitro-5-(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD09701814
  • Cas Number : 1170852-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart