PRI424056854 Princeton BioMolecular Research, Inc.

1-[4-(1-ethyl-3-methyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidin-2-yl]piperidin-4-amine

  • MDL number MFCD09701829
  • Cas Number : 1170525-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart