PRI424057095 Princeton BioMolecular Research, Inc.

1-ethyl-5-pyrrolidin-2-yl-1H-pyrazole

  • MDL number MFCD09701946
  • Cas Number : 1171665-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart