PRI424057099 Princeton BioMolecular Research, Inc.

1-isobutyl-5-pyrrolidin-2-yl-1H-pyrazole

  • MDL number MFCD09701950
  • Cas Number : 1172803-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart