PRI424057104 Princeton BioMolecular Research, Inc.

3-isopropyl-5-pyrrolidin-2-ylisoxazole

  • MDL number MFCD09701954
  • Cas Number : 1018163-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart