PRI424057226 Princeton BioMolecular Research, Inc.

2-(2,2-difluoroethyl)-3,4-dimethyl-2,7-dihydro-6H-pyrazolo[3,4-b]pyridin-6-one

  • MDL number MFCD09834178
  • Cas Number : 1018163-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart