PRI424057250 Princeton BioMolecular Research, Inc.

[2-(1-ethyl-1H-pyrazol-4-yl)-1H-benzimidazol-1-yl]acetic acid

  • MDL number MFCD09834204
  • Cas Number : 1172275-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart