PRI424057268 Princeton BioMolecular Research, Inc.

3-[4-(difluoromethyl)-6-oxo-2-propyl-2,6-dihydro-7H-pyrazolo[3,4-b]pyridin-7-yl]propanoic acid

  • MDL number MFCD09834222
  • Cas Number : 1018126-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart