PRI424057271 Princeton BioMolecular Research, Inc.

3-[4-(difluoromethyl)-2-isopropyl-6-oxo-2,6-dihydro-7H-pyrazolo[3,4-b]pyridin-7-yl]propanoic acid

  • MDL number MFCD09834225
  • Cas Number : 1018164-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart