PRI424057280 Princeton BioMolecular Research, Inc.

3-{[[(1-ethyl-1H-pyrazol-5-yl)methyl](methyl)amino]sulfonyl}thiophene-2-carboxylic acid

  • MDL number MFCD09834234
  • Cas Number : 1171417-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart