PRI424057323 Princeton BioMolecular Research, Inc.

2-(2,2-difluoroethyl)-3-methyl-4-(trifluoromethyl)-2,7-dihydro-6H-pyrazolo[3,4-b]pyridin-6-one

  • MDL number MFCD09834270
  • Cas Number : 1018127-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart