PRI424057338 Princeton BioMolecular Research, Inc.

6-chloro-4-(difluoromethyl)-2-isopropyl-2H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD09834289
  • Cas Number : 1018127-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart