PRI424057344 Princeton BioMolecular Research, Inc.

2-butyl-6-chloro-4-(difluoromethyl)-3-methyl-2H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD09834294
  • Cas Number : 1018127-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart