PRI424057354 Princeton BioMolecular Research, Inc.

6-chloro-2-(2,2-difluoroethyl)-3,4-dimethyl-2H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD09834305
  • Cas Number : 1018127-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart