PRI424057383 Princeton BioMolecular Research, Inc.

[2-(4-methyl-1H-pyrazol-1-yl)butyl]amine

  • MDL number MFCD10001531
  • Cas Number : 1171366-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart