PRI424057450 Princeton BioMolecular Research, Inc.

5-(4-tert-butylphenyl)-4,5-dihydroisoxazole-3-carboxylic acid

  • MDL number MFCD09859238
  • Cas Number : 1018046-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart