PRI424057496 Princeton BioMolecular Research, Inc.

1-(1-pyrrolidin-2-ylethyl)-1H-pyrazole

  • MDL number MFCD09859277
  • Cas Number : 1172941-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart