PRI424057531 Princeton BioMolecular Research, Inc.

methyl 4-(1-methyl-1H-pyrazol-3-yl)-2,4-dioxobutanoate

  • MDL number MFCD09859307
  • Cas Number : 875553-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart