PRI424057556 Princeton BioMolecular Research, Inc.

6-chloro-2-cyclopentyl-4-(difluoromethyl)-2H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD09859331
  • Cas Number : 1018166-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart