PRI424057562 Princeton BioMolecular Research, Inc.

1-[(5-isopropylisoxazol-3-yl)methyl]piperazine

  • MDL number MFCD09859413
  • Cas Number : 1018167-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart