PRI424057640 Princeton BioMolecular Research, Inc.

1-ethyl-3-(5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyridin-3-yl)-1H-pyrazol-4-amine

  • MDL number MFCD09859610
  • Cas Number : 1170155-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart