PRI424057644 Princeton BioMolecular Research, Inc.

3-cyclopropyl-1-ethyl-1H-pyrazole-5-carbonitrile

  • MDL number MFCD10001537
  • Cas Number : 1171385-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart