PRI424057648 Princeton BioMolecular Research, Inc.

5-(4-methylphenyl)-4,5-dihydroisoxazole-3-carboxylic acid

  • MDL number MFCD10001543
  • Cas Number : 1018170-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart