PRI424057653 Princeton BioMolecular Research, Inc.

ethyl 4-(difluoromethyl)-1,2,3-thiadiazole-5-carboxylate

  • MDL number MFCD10001548
  • Cas Number : 1018170-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart