PRI424057663 Princeton BioMolecular Research, Inc.

{3-[2-(4-chlorophenyl)pyrrolidin-1-yl]propyl}amine

  • MDL number MFCD10001557
  • Cas Number : 1018051-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart