PRI424057669 Princeton BioMolecular Research, Inc.

[(1,5-diethyl-1H-pyrazol-4-yl)methyl]amine

  • MDL number MFCD08700603
  • Cas Number : 1007540-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart