PRI424057674 Princeton BioMolecular Research, Inc.

6-chloro-2,3,4-trimethyl-2H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD10001562
  • Cas Number : 1018170-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart