PRI424057695 Princeton BioMolecular Research, Inc.

{2-[(3-cyclopropyl-1-methyl-1H-pyrazol-5-yl)methoxy]ethyl}amine

  • MDL number MFCD10001579
  • Cas Number : 1171355-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart