PRI424057705 Princeton BioMolecular Research, Inc.

3-[5-cyclopropyl-7-(difluoromethyl)[1,2,4]triazolo[1,5-a]pyrimidin-2-yl]propanoic acid

  • MDL number MFCD10001596
  • Cas Number : 1018052-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart