PRI424057812 Princeton BioMolecular Research, Inc.

1-isopropyl-3-(trifluoromethyl)-1H-pyrazol-5-amine

  • MDL number MFCD10699028
  • Cas Number : 1171326-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart