PRI424057829 Princeton BioMolecular Research, Inc.

[1-(1-methyl-1H-pyrazol-4-yl)-2-morpholin-4-ylethyl]amine

  • MDL number MFCD11215316
  • Cas Number : 1174880-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart