PRI424057836 Princeton BioMolecular Research, Inc.

[7-(1,5-dimethyl-1H-pyrazol-4-yl)[1,2,4]triazolo[1,5-a]pyrimidin-2-yl]acetic acid

  • MDL number MFCD11215343
  • Cas Number : 1174835-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart