PRI424057853 Princeton BioMolecular Research, Inc.

{[5-(1-methyl-1H-pyrazol-4-yl)isoxazol-3-yl]methyl}amine

  • MDL number MFCD08700589
  • Cas Number : 1007540-25-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart