PRI424057880 Princeton BioMolecular Research, Inc.

2,2-difluoro-1-(1-methyl-1H-pyrazol-4-yl)ethanone

  • MDL number MFCD10699051
  • Cas Number : 1174839-82-7
  • Molecular Weight : 160.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,2-difluoro-1-(1-methyl-1H-pyrazol-4-yl)ethanone
Inchi Code 1S/C6H6F2N2O/c1-10-3-4(2-9-10)5(11)6(7)8/h2-3,6H,1H3
Inchi Key VBEPRYRQRBOKGI-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart