PRI424057883 Princeton BioMolecular Research, Inc.

1-(1-ethyl-1H-pyrazol-4-yl)-2,2-difluoroethanone

  • MDL number MFCD10699054
  • Cas Number : 1174879-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart