PRI424057898 Princeton BioMolecular Research, Inc.

3-(ethoxycarbonyl)-1-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD10699065
  • Cas Number : 1174830-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart