PRI424057921 Princeton BioMolecular Research, Inc.

1-(5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyridin-3-ylmethyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD10699085
  • Cas Number : 1174854-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart