PRI424057939 Princeton BioMolecular Research, Inc.

[1-(2,2,2-trifluoroethyl)piperidin-4-yl]acetic acid

  • MDL number MFCD11215377
  • Cas Number : 1159983-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart