PRI424057948 Princeton BioMolecular Research, Inc.

7-(difluoromethyl)[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylic acid

  • MDL number MFCD11215381
  • Cas Number : 1160246-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart