PRI424057950 Princeton BioMolecular Research, Inc.

2-(2,2-difluoroethyl)-3-methyl-6-oxo-6,7-dihydro-2H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD12405242
  • Cas Number : 1160246-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart