PRI424058056 Princeton BioMolecular Research, Inc.

1-propyl-4-(6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepin-3-yl)-1H-pyrazol-3-amine

  • MDL number MFCD12405273
  • Cas Number : 1174856-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart