PRI424058072 Princeton BioMolecular Research, Inc.

1-ethyl-5-(trifluoromethyl)-1H-pyrazol-3-amine

  • MDL number MFCD09701931
  • Cas Number : 1365988-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart