PRI424058147 Princeton BioMolecular Research, Inc.

1-(6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-3-ylmethyl)-5-methyl-1H-pyrazol-3-amine

  • MDL number MFCD12548019
  • Cas Number : 1174871-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart