PRI424058181 Princeton BioMolecular Research, Inc.

5-[(4-aminopiperidin-1-yl)methyl]thiophene-2-carboxylic acid

  • MDL number MFCD12548043
  • Cas Number : 1174863-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart