PRI424058196 Princeton BioMolecular Research, Inc.

3-(difluoromethyl)-1-methyl-1H-pyrazol-5-amine

  • MDL number MFCD15976250
  • Cas Number : 1245806-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart