PRI424058436 Princeton BioMolecular Research, Inc.

N-(2,2-difluoroethyl)-N-(tetrahydrofuran-2-ylmethyl)amine

  • MDL number MFCD12772330
  • Cas Number : 1182935-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart