PRI424058448 Princeton BioMolecular Research, Inc.

N-(2,2-difluoroethyl)-N-(2-methoxyethyl)amine

  • MDL number MFCD12807384
  • Cas Number : 1178929-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart