PRI424058461 Princeton BioMolecular Research, Inc.

4-chloro-1-cyclopentyl-1H-pyrazole-5-sulfonyl chloride

  • MDL number MFCD15976282
  • Cas Number : 1245808-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart